missing translation for 'onlineSavingsMsg'
Learn More

D-Methionine methyl ester hydrochloride, 98%

CAS: 69630-60-0 | C6H14ClNO2S | 199.69 g/mol

Supplier:  Thermo Scientific Chemicals H6342214

 View more versions of this product

Catalog No. AAH6342214


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D-Methionine methyl ester hydrochloride
C6H14ClNO2S
h-d-met-ome.hcl, d-methionine methyl ester hydrochloride, h-d-met-ome hcl, methyl d-methioninate-hydrogen chloride 1/1, methyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, h-d-met-omecl, d-methionine methyl ester hcl, methyl d-methioninate hydrochloride, d methionine methyl ester hydrochloride
Cl.COC(=O)[C@H](N)CCSC
25 g
12888616
98%
69630-60-0
MFCD00070385
MEVUPUNLVKELNV-NUBCRITNSA-N
methyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride
199.69
199.7
Solid

Chemical Identifiers

69630-60-0
199.69
MEVUPUNLVKELNV-NUBCRITNSA-N
12888616
Cl.COC(=O)[C@H](N)CCSC
C6H14ClNO2S
MFCD00070385
h-d-met-ome.hcl, d-methionine methyl ester hydrochloride, h-d-met-ome hcl, methyl d-methioninate-hydrogen chloride 1/1, methyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, h-d-met-omecl, d-methionine methyl ester hcl, methyl d-methioninate hydrochloride, d methionine methyl ester hydrochloride
methyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only