missing translation for 'onlineSavingsMsg'
Learn More

D-Methionine methyl ester hydrochloride, 98%

Catalog No. p-7054964
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6342206 5 g
AAH6342214 25 g
AAH6342222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6342206 Supplier Thermo Scientific Chemicals Supplier No. H6342206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69630-60-0 | C6H14ClNO2S | 199.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69630-60-0
Molecular Formula C6H14ClNO2S
Molecular Weight (g/mol) 199.69
MDL Number MFCD00070385
InChI Key MEVUPUNLVKELNV-NUBCRITNSA-N
Synonym h-d-met-ome.hcl, d-methionine methyl ester hydrochloride, h-d-met-ome hcl, methyl d-methioninate-hydrogen chloride 1/1, methyl 2r-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, h-d-met-omecl, d-methionine methyl ester hcl, methyl d-methioninate hydrochloride, d methionine methyl ester hydrochloride
PubChem CID 12888616
IUPAC Name methyl (2R)-2-amino-4-methylsulfanylbutanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](N)CCSC

Specifications

Quantity 5 g
Formula Weight 199.7
Percent Purity 98%
Physical Form Solid
Chemical Name or Material D-Methionine methyl ester hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only