accessibility menu, dialog, popup

UserName

CH3OO(R)SH3CNH2HCl

CAS RN 69630-60-0

IUPAC Name: methyl (2R)-2-amino-4-(methylsulfanyl)butanoate hydrochloride
Synonyms: methyl (2R)-2-amino-4-(methylsulfanyl)butanoate hydrochloride|H-D-MET-OME HCL
Molecular Weight (g/mol): 199.69
Molecular Formula: C6H14ClNO2S
InChi Key: MEVUPUNLVKELNV-NUBCRITNSA-N
SMILES: Cl.COC(=O)[C@H](N)CCSC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)