Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ D-Methionine, 99+%
$83.42 - $626.60
Chemical Identifiers
| CAS | 348-67-4 |
|---|---|
| Molecular Formula | C5H11NO2S |
| Molecular Weight (g/mol) | 149.208 |
| MDL Number | MFCD00002622 |
| InChI Key | FFEARJCKVFRZRR-SCSAIBSYSA-N |
| Synonym | d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid |
| PubChem CID | 84815 |
| ChEBI | CHEBI:16867 |
| IUPAC Name | (2R)-2-amino-4-methylsulfanylbutanoic acid |
| SMILES | CSCCC(C(=O)O)N |
| missing translation for 'productCode' | missing translation for 'brand' | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| missing translation for 'productCode' | missing translation for 'brand' | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC227210050
|
Thermo Scientific™
227210050 |
5g | Glass bottle |
Each for $83.42
|
|
||||
|
AC227210250
|
Thermo Scientific™
227210250 |
25g | Glass bottle |
Each for $239.15
|
|
||||
|
AC227211000
|
Thermo Scientific™
227211000 |
100g | Glass bottle |
Each for $626.60
|
|
||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C5H11NO2S | |
| MFCD00002622 | |
| d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid | |
| CHEBI:16867 | |
| CSCCC(C(=O)O)N |
Specifications
| 200ppm max. | |
| 1ppm max. | |
| 10ppm max. | |
| 10ppm max. | |
| 99+% | |
| C5H11NO2S | |
| MFCD00002622 | |
| 300ppm max. | |
| d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid | |
| FFEARJCKVFRZRR-SCSAIBSYSA-N | |
| − 24.00 (20.00°C c=8,6N HCl) | |
| 149.208 | |
| CHEBI:16867 | |
| ≥99% |
| 348-67-4 | |
| 300ppm max. | |
| Authentic | |
| 0.3% max. | |
| (TLC) not detectable | |
| CH3SCH2CH2CH(NH2)CO2H | |
| − 24.00 | |
| 98% min. (c=3, H2O, 430nm) 1cm cell | |
| 0.1% max. | |
| CSCCC(C(=O)O)N | |
| (2R)-2-amino-4-methylsulfanylbutanoic acid | |
| 84815 | |
| 149.21 | |
| D-Methionine, 99+% |
Safety and Handling
EINECSNumber : 206-483-6
RTECSNumber : PD0455000
TSCA : TSCA
RUO – Research Use Only