missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Methionine, 99+%

Catalog No. AC227210050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227210050 5 g Glass Bottle
AC227210250 25 g Glass Bottle
AC227211000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC227210050 Supplier Thermo Scientific Chemicals Supplier No. 227210050

CAS: 348-67-4 | C5H11NO2S | 149.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 348-67-4
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00002622
InChI Key FFEARJCKVFRZRR-SCSAIBSYSA-N
Synonym d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid
PubChem CID 84815
ChEBI CHEBI:16867
SMILES CSCC[C@@H](N)C(O)=O

Specifications

Melting Point 273.0°C to 275.0°C
Ammonium 200ppm max.
Arsenic (As) 1ppm max.
Color White
Chlorine (Cl) 300ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 0.3% max.
Assay Percent Range 99+%
Other Amino Acids (TLC) not detectable
Packaging Glass Bottle
Linear Formula CH3SCH2CH2CH(NH2)CO2H
Specific Rotation − 24.00
Sulfate 300ppm max.
Transmittance 98% min. (c=3, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: soluble in water: 53g/L (20°C).
IUPAC Name (2R)-2-amino-4-(methylsulfanyl)butanoic acid
Specific Rotation Condition − 24.00 (20.00°C c=8,6N HCl)
Quantity 5 g
Formula Weight 149.21
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material D-Methionine, 99+%
Show More Show Less
EINECSNumber 206-483-6
RTECSNumber PD0455000
TSCA TSCA

RUO – Research Use Only