missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ D-Methionine, 99+%
Supplier: Thermo Scientific™ 227210250
| Quantity | 25g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C5H11NO2S | |
| MFCD00002622 | |
| d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid | |
| CHEBI:16867 | |
| CSCCC(C(=O)O)N |
Specifications
| D-Methionine | |
| 200ppm max. | |
| 1ppm max. | |
| 10ppm max. | |
| 10ppm max. | |
| 99+% | |
| C5H11NO2S | |
| CH3SCH2CH2CH(NH2)CO2H | |
| − 24.00 | |
| 98% min. (c=3, H2O, 430nm) 1cm cell | |
| 0.1% max. | |
| CSCCC(C(=O)O)N | |
| (2R)-2-amino-4-methylsulfanylbutanoic acid | |
| 25g | |
| CHEBI:16867 | |
| ≥99% |
| 99+% | |
| 348-67-4 | |
| 300ppm max. | |
| Authentic | |
| 0.3% max. | |
| (TLC) not detectable | |
| Glass bottle | |
| MFCD00002622 | |
| 300ppm max. | |
| d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid | |
| FFEARJCKVFRZRR-SCSAIBSYSA-N | |
| − 24.00 (20.00°C c=8,6N HCl) | |
| 149.208 | |
| 84815 | |
| 149.21 |
Safety and Handling
EINECSNumber : 206-483-6
RTECSNumber : PD0455000
TSCA : TSCA