missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Mannitol 99.0+%, TCI America™
$43.67 - $83.54
Chemical Identifiers
| CAS | 69-65-8 |
|---|---|
| Molecular Formula | C6H14O6 |
| Molecular Weight (g/mol) | 182.17 |
| MDL Number | MFCD00064287 |
| InChI Key | FBPFZTCFMRRESA-UHFFFAOYNA-N |
| Synonym | d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex |
| PubChem CID | 6251 |
| ChEBI | CHEBI:16899 |
| IUPAC Name | hexane-1,2,3,4,5,6-hexol |
| SMILES | OCC(O)C(O)C(O)C(O)CO |
Chemical Identifiers
| C6H14O6 | |
| MFCD00064287 | |
| d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex | |
| CHEBI:16899 | |
| OCC(O)C(O)C(O)C(O)CO |
Specifications
| 69-65-8 | |
| White | |
| MFCD00064287 | |
| FBPFZTCFMRRESA-UHFFFAOYNA-N | |
| hexane-1,2,3,4,5,6-hexol | |
| 6251 | |
| 182.17 | |
| Crystalline Powder |
| 168°C | |
| C6H14O6 | |
| d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex | |
| OCC(O)C(O)C(O)C(O)CO | |
| 182.17 | |
| CHEBI:16899 | |
| ≥99.0% (T) | |
| D-Mannitol |
Safety and Handling
EINECSNumber : (8)-0049&(9)-1375
RTECSNumber : OP2060000
TSCA : Yes