Learn More
D-Mannitol (Powder/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical M120500
Specifications
| Mannitol | |
| 69-65-8 | |
| White | |
| 0.01% max. | |
| Poly Bottle | |
| MFCD00064287 | |
| 23.3 to 24.3° (+ or -) | |
| 0.01% max. Insoluble Matter | |
| OCC(O)C(O)C(O)C(O)CO | |
| 182.17 | |
| CHEBI:16899 | |
| Certified ACS |
| D-Mannitol | |
| 165°C | |
| 295°C | |
| 0.05% max. | |
| C6H14O6 | |
| 500 g | |
| d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex | |
| FBPFZTCFMRRESA-UHFFFAOYNA-N | |
| hexane-1,2,3,4,5,6-hexol | |
| 6251 | |
| 182.17 | |
| Solid |
Chemical Identifiers
| 69-65-8 | |
| 182.17 | |
| FBPFZTCFMRRESA-UHFFFAOYNA-N | |
| 6251 | |
| hexane-1,2,3,4,5,6-hexol |
| C6H14O6 | |
| MFCD00064287 | |
| d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex | |
| CHEBI:16899 | |
| OCC(O)C(O)C(O)C(O)CO |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Handle in accordance with good industrial hygiene and safety practice. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:1
Instability:0