missing translation for 'onlineSavingsMsg'
Learn More

D-Mannitol (Powder/Certified ACS), Fisher Chemical™

Catalog No. p-154985
Change view
Click to view available options
Quantity:
500 g
3 kg
60 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M120500 500 g Poly Bottle
M1203 3 kg Poly Pail
M12060 60 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M120500 Supplier Thermo Fisher Scientific Supplier No. M120 500
Only null left

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Alpha Vector Mannitol
Melting Point 165°C
Color White
Boiling Point 295°C
Ignition Residue 0.01% max.
Loss on Drying 0.05% max.
Packaging Poly Bottle
Quantity 500 g
Specific Rotation 23.3 to 24.3° (+ or -)
Solubility Information 0.01% max. Insoluble Matter
Formula Weight 182.17
Grade Certified ACS
Physical Form Solid
Chemical Name or Material D-Mannitol
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Handle in accordance with good industrial hygiene and safety practice. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:0