Learn More
Thermo Scientific Chemicals D-Mannitol, ACS
CAS: 69-65-8 | C6H14O6 | 182.17 g/mol
$146.75 - $440.47
Chemical Identifiers
| CAS | 69-65-8 |
|---|---|
| Molecular Formula | C6H14O6 |
| Molecular Weight (g/mol) | 182.17 |
| MDL Number | MFCD00064287 |
| InChI Key | FBPFZTCFMRRESA-UHFFFAOYNA-N |
| Synonym | d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex |
| PubChem CID | 6251 |
| ChEBI | CHEBI:16899 |
| IUPAC Name | (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol |
| SMILES | OCC(O)C(O)C(O)C(O)CO |
Description
D-Mannitol is used with boric acid to make dry electrolytic condensers for radio applications, artificial resins and plasticizers. It is used as excipient for chewable tablets due to its pleasant taste. It is used in volumetric analysis of boric acid in analytical chemistry. It is also used in the food industry as anticaking, free-flow agent, flavoring agent, stabilizer, thickener and nutritive sweetener. Further, it is used in medicine as diuretic and in the determination of renal function. In addition, it is used as a formulation aid, lubricant, non-nutritive sweetener and solvent for foods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C6H14O6 | |
| MFCD00064287 | |
| d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex | |
| CHEBI:16899 | |
| OCC(O)C(O)C(O)C(O)CO |
Specifications
| 69-65-8 | |
| 1.489 g/mL | |
| C6H14O6 | |
| 1721898 | |
| d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex | |
| FBPFZTCFMRRESA-UHFFFAOYNA-N | |
| (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol | |
| 6251 | |
| 182.17 | |
| Crystalline |
| 165°C to 167°C | |
| 290°C to 295°C (3.5 mm) | |
| MFCD00064287 | |
| 14,5745 | |
| Soluble in water,alcohol,pyridine,aniline,glycerol and aqueous solution of alkalis; Insoluble in ether. | |
| OCC(O)C(O)C(O)C(O)CO | |
| 182.17 | |
| CHEBI:16899 | |
| ACS | |
| D-Mannitol, For ACS analysis |
Safety and Handling
EINECSNumber : 200-711-8
RTECSNumber : OP2060000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only