missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Mannitol, ACS

Catalog No. AA3334236
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3334236 500 g
AA33342A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3334236 Supplier Thermo Scientific Chemicals Supplier No. 03334236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 69-65-8 | C6H14O6 | 182.17 g/mol

D-Mannitol is used with boric acid to make dry electrolytic condensers for radio applications, artificial resins and plasticizers. It is used as excipient for chewable tablets due to its pleasant taste. It is used in volumetric analysis of boric acid in analytical chemistry. It is also used in the food industry as anticaking, free-flow agent, flavoring agent, stabilizer, thickener and nutritive sweetener. Further, it is used in medicine as diuretic and in the determination of renal function. In addition, it is used as a formulation aid, lubricant, non-nutritive sweetener and solvent for foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
IUPAC Name (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Melting Point 165°C to 167°C
Density 1.489 g/mL
Boiling Point 290°C to 295°C (3.5 mm)
Quantity 500 g
Beilstein 1721898
Merck Index 14,5745
Solubility Information Soluble in water,alcohol,pyridine,aniline,glycerol and aqueous solution of alkalis; Insoluble in ether.
Formula Weight 182.17
Grade ACS
Physical Form Crystalline
Chemical Name or Material D-Mannitol, For ACS analysis
Show More Show Less
EINECSNumber 200-711-8
RTECSNumber OP2060000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only