missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(-)-Lyxose 98.0+%, TCI America™
$132.90 - $748.05
Chemical Identifiers
| CAS | 1114-34-7 |
|---|---|
| Molecular Formula | C5H10O5 |
| Molecular Weight (g/mol) | 150.13 |
| MDL Number | MFCD00064362 |
| InChI Key | PYMYPHUHKUWMLA-UOWFLXDJSA-N |
| Synonym | 2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci |
| PubChem CID | 65550 |
| ChEBI | CHEBI:16789 |
| IUPAC Name | (2S,3S,4R)-2,3,4,5-tetrahydroxypentanal |
| SMILES | C(C(C(C(C=O)O)O)O)O |
Chemical Identifiers
| 1114-34-7 | |
| 150.13 | |
| PYMYPHUHKUWMLA-UOWFLXDJSA-N | |
| 65550 | |
| (2S,3S,4R)-2,3,4,5-tetrahydroxypentanal |
| C5H10O5 | |
| MFCD00064362 | |
| 2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci | |
| CHEBI:16789 | |
| C(C(C(C(C=O)O)O)O)O |
Specifications
| 1114-34-7 | |
| C5H10O5 | |
| 1 g | |
| PYMYPHUHKUWMLA-UOWFLXDJSA-N | |
| (2S,3S,4R)-2,3,4,5-tetrahydroxypentanal | |
| 65550 | |
| 150.13 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00064362 | |
| 2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci | |
| C(C(C(C(C=O)O)O)O)O | |
| 150.13 | |
| CHEBI:16789 | |
| ≥98.0% (HPLC) | |
| D-(-)-Lyxose |
Safety and Handling
TSCA : Yes