missing translation for 'onlineSavingsMsg'
Learn More

D-Lyxose, 99%

Catalog No. AAA1700706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1700706 5 g
AAA1700714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1700706 Supplier Thermo Scientific Chemicals Supplier No. A1700706

CAS: 1114-34-7 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1114-34-7
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00064362
InChI Key PYMYPHUHKUWMLA-UOWFLXDJSA-N
Synonym 2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci
PubChem CID 65550
ChEBI CHEBI:16789
IUPAC Name (2S,3S,4R)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O

Specifications

Melting Point 109°C to 115°C
Density 1.545
Quantity 5 g
Beilstein 1723083
Sensitivity Hygroscopic
Merck Index 14,5641
Formula Weight 150.13
Percent Purity 99%
Chemical Name or Material D-Lyxose
EINECSNumber 214-212-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only