missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(-)-Lysine Monohydrochloride 98.0+%, TCI America™
$50.05 - $148.25
Chemical Identifiers
| CAS | 7274-88-6 |
|---|---|
| Molecular Formula | C6H15ClN2O2 |
| Molecular Weight (g/mol) | 182.65 |
| MDL Number | MFCD00012920 |
| InChI Key | BVHLGVCQOALMSV-XZNNNFJINA-N |
| Synonym | d-lysine hydrochloride, d-lysine monohydrochloride, d-lysine hcl, d-lysine, monohydrochloride, h-d-lys-oh.hcl, d-lysine, hydrochloride, unii-kk611jde1n, lysine, monohydrochloride, d, r-2,6-diaminohexanoic acid hydrochloride, lysine, hydrochloride, d |
| PubChem CID | 81691 |
| ChEBI | CHEBI:53558 |
| IUPAC Name | (2R)-2,6-diaminohexanoic acid hydrochloride |
| SMILES | Cl.NCCCC[C@@H](N)C(O)=O |
Chemical Identifiers
| 7274-88-6 | |
| 182.65 | |
| BVHLGVCQOALMSV-XZNNNFJINA-N | |
| 81691 | |
| (2R)-2,6-diaminohexanoic acid hydrochloride |
| C6H15ClN2O2 | |
| MFCD00012920 | |
| d-lysine hydrochloride, d-lysine monohydrochloride, d-lysine hcl, d-lysine, monohydrochloride, h-d-lys-oh.hcl, d-lysine, hydrochloride, unii-kk611jde1n, lysine, monohydrochloride, d, r-2,6-diaminohexanoic acid hydrochloride, lysine, hydrochloride, d | |
| CHEBI:53558 | |
| Cl.NCCCC[C@@H](N)C(O)=O |
Specifications
| 7274-88-6 | |
| C6H15ClN2O2 | |
| 5 g | |
| BVHLGVCQOALMSV-XZNNNFJINA-N | |
| (2R)-2,6-diaminohexanoic acid hydrochloride | |
| 81691 | |
| 182.65 | |
| Crystalline Powder |
| White | |
| MFCD00012920 | |
| d-lysine hydrochloride, d-lysine monohydrochloride, d-lysine hcl, d-lysine, monohydrochloride, h-d-lys-oh.hcl, d-lysine, hydrochloride, unii-kk611jde1n, lysine, monohydrochloride, d, r-2,6-diaminohexanoic acid hydrochloride, lysine, hydrochloride, d | |
| Cl.NCCCC[C@@H](N)C(O)=O | |
| 182.65 | |
| CHEBI:53558 | |
| ≥98.0% (T) | |
| D-(-)-Lysine Monohydrochloride |
Safety and Handling
EINECSNumber : (2)-3200
RTECSNumber : OL5632500
TSCA : Yes