missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(-)-Lysine Monohydrochloride 98.0+%, TCI America™
Supplier: TCI America L012825G
Specifications
| D-(-)-Lysine Monohydrochloride | |
| White | |
| MFCD00012920 | |
| d-lysine hydrochloride, d-lysine monohydrochloride, d-lysine hcl, d-lysine, monohydrochloride, h-d-lys-oh.hcl, d-lysine, hydrochloride, unii-kk611jde1n, lysine, monohydrochloride, d, r-2,6-diaminohexanoic acid hydrochloride, lysine, hydrochloride, d | |
| Cl.NCCCC[C@@H](N)C(O)=O | |
| 182.65 | |
| CHEBI:53558 | |
| ≥98.0% (T) |
| 7274-88-6 | |
| C6H15ClN2O2 | |
| 25 g | |
| BVHLGVCQOALMSV-XZNNNFJINA-N | |
| (2R)-2,6-diaminohexanoic acid hydrochloride | |
| 81691 | |
| 182.65 | |
| Crystalline Powder |
Chemical Identifiers
| 7274-88-6 | |
| 182.65 | |
| BVHLGVCQOALMSV-XZNNNFJINA-N | |
| 81691 | |
| (2R)-2,6-diaminohexanoic acid hydrochloride |
| C6H15ClN2O2 | |
| MFCD00012920 | |
| d-lysine hydrochloride, d-lysine monohydrochloride, d-lysine hcl, d-lysine, monohydrochloride, h-d-lys-oh.hcl, d-lysine, hydrochloride, unii-kk611jde1n, lysine, monohydrochloride, d, r-2,6-diaminohexanoic acid hydrochloride, lysine, hydrochloride, d | |
| CHEBI:53558 | |
| Cl.NCCCC[C@@H](N)C(O)=O |
Safety and Handling
EINECSNumber : (2)-3200
RTECSNumber : OL5632500
TSCA : Yes