missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ D-Luciferin sodium salt

Catalog No. 542750
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
542750 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 542750 Supplier Tocris Bioscience™ Supplier No. 5427/50

Firefly luciferase substrate

D-Luciferin sodium salt is a firefly luciferase substrate (Km = ∽2 μM). Used in many standard bioluminescence imaging (BLI) techniques, including monitoring tumour growth, and investigating intracellular signaling activity in vitro and in vivo. Cell permeable. Emission maximum = 562 nm.

Specifications

Quantity 50 mg
Molecular Formula C11H7N2NaO3S2
Product Type D-Luciferin sodium salt
Purity >99%
Molecular Weight (g/mol) 302.3
Inhibitors D-Luciferin sodium salt

For Research Use Only