missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ D-Leucine, 99%

$84.78 - $247.24

Chemical Identifiers

CAS 328-38-1
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD00063088
InChI Key ROHFNLRQFUQHCH-RXMQYKEDSA-N
Synonym d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin
PubChem CID 439524
ChEBI CHEBI:28225
IUPAC Name (2R)-2-amino-4-methylpentanoic acid
SMILES CC(C)CC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC226900025
View Documents Promotion Details
Thermo Scientific™
226900025
2.5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC226900100
View Documents Promotion Details
Thermo Scientific™
226900100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

328-38-1
131.175
ROHFNLRQFUQHCH-RXMQYKEDSA-N
439524
(2R)-2-amino-4-methylpentanoic acid
C6H13NO2
MFCD00063088
d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin
CHEBI:28225
CC(C)CC(C(=O)O)N

Specifications

328-38-1
10ppm max.
10ppm max.
99%
(CH3)2CHCH2CH(NH2)CO2H
0.1% max.
-14.9° to -16° (20°C, 589nm) (c=4, 6N HCl)
ROHFNLRQFUQHCH-RXMQYKEDSA-N
(2R)-2-amino-4-methylpentanoic acid
131.175
CHEBI:28225
≥98.5%
2ppm max.
Authentic
0.2% max.
C6H13NO2
MFCD00063088
04, 446
d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin
CC(C)CC(C(=O)O)N
−15.45° (20°C c=4,6N HCl)
439524
131.17
D-Leucine, 99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 206-327-7

RTECSNumber : OH2840000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only