accessibility menu, dialog, popup

UserName

CAS RN 328-38-1

IUPAC Name: (2R)-2-amino-4-methylpentanoic acid
Synonyms: D-leucine|L-(+)-leucine|leucine, L-|1-leucine|leucina|leucine,d|leucinum|α-aminoisocaproic acid|L-a-aminoisocaproic acid|L isomer leucine
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: ROHFNLRQFUQHCH-RXMQYKEDSA-N
SMILES: CC(C)C[C@@H](N)C(O)=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)