missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Leucine, 99%

Catalog No. AC226900025
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226900025 2.5 g Glass Bottle
AC226900100 10 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226900025 Supplier Thermo Scientific Chemicals Supplier No. 226900025

CAS: 328-38-1 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 328-38-1
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00063088
InChI Key ROHFNLRQFUQHCH-RXMQYKEDSA-N
Synonym d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin
PubChem CID 439524
ChEBI CHEBI:28225
IUPAC Name (2R)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@@H](N)C(O)=O

Specifications

Arsenic (As) 2ppm max.
Color White
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 0.2% max.
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2CH(NH2)CO2H
Residue after Ignition 0.1% max.
Beilstein 04, 446
Specific Rotation -14.9° to -16° (20°C, 589nm) (c=4, 6N HCl)
Specific Rotation Condition −15.45° (20°C c=4,6N HCl)
Quantity 2.5 g
Formula Weight 131.17
Percent Purity ≥98.5%
Physical Form Crystalline Powder, Crystals, or Flakes
Chemical Name or Material D-Leucine, 99%
Show More Show Less
EINECSNumber 206-327-7
RTECSNumber OH2840000
TSCA TSCA

RUO – Research Use Only