missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals α-D-Lactose monohydrate, ACS

μa-D-Lactose monohydrate, ACS, CAS # 64044-51-5, is the crystalline form of D-Lactose, which is the major sugar found in milk. | CAS: 5989-81-1 | C12H24O12 | 360.312 g/mol

Supplier:  Thermo Scientific Chemicals 03621836

 View more versions of this product

Catalog No. AA3621836


Only null left
Explore available promotions

Description

Description

It is also heavily used in the pharmaceutical industry as a pill filler. They are capable of fermenting lactose for ethanol production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• μa-D-Lactose monohydrate, ACS is the crystalline form of D-Lactose
• It is composed of galactose and glucose subunits

Application

• μa-D-Lactose monohydrate can be used to ferment lactose for ethanol production in laboratory settings
• It may be used as a reference standard for lactose quantitation

Specifications

Specifications

α-D-Lactose monohydrate
For analysis ACS
Crystalline
C12H24O12
3768231
d-lactose monohydrate, d-+-lactosemonohydrate
WSVLPVUVIUVCRA-ZEAJSSQNSA-N
(3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
133126686
ACS
∽218°C (decomposition)
5989-81-1
500 g
MFCD00150747
14,5343
Soluble in water (warm) (0.1g/mL),and ethanol (very slightly soluble). Insoluble in chloroform,and ether.
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
360.312
360.32 (342.30 Anhydrous)
1.525 g/mL

Chemical Identifiers

5989-81-1
360.312
WSVLPVUVIUVCRA-ZEAJSSQNSA-N
133126686
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
C12H24O12
MFCD00150747
d-lactose monohydrate, d-+-lactosemonohydrate
(3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-559-2

RTECSNumber : OD9625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only