missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals α-D-Lactose monohydrate, ACS

Catalog No. p-7050223
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3621836 500 g
AA36218A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3621836 Supplier Thermo Scientific Chemicals Supplier No. 03621836
Only null left

μa-D-Lactose monohydrate, ACS, CAS # 64044-51-5, is the crystalline form of D-Lactose, which is the major sugar found in milk. | CAS: 5989-81-1 | C12H24O12 | 360.312 g/mol

It is also heavily used in the pharmaceutical industry as a pill filler. They are capable of fermenting lactose for ethanol production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• μa-D-Lactose monohydrate, ACS is the crystalline form of D-Lactose
• It is composed of galactose and glucose subunits

Application

• μa-D-Lactose monohydrate can be used to ferment lactose for ethanol production in laboratory settings
• It may be used as a reference standard for lactose quantitation

Chemical Identifiers

CAS 5989-81-1
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.312
MDL Number MFCD00150747
InChI Key WSVLPVUVIUVCRA-ZEAJSSQNSA-N
Synonym d-lactose monohydrate, d-+-lactosemonohydrate
PubChem CID 133126686
IUPAC Name (3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O

Specifications

Melting Point ∽218°C (decomposition)
Physical Form Crystalline
Quantity 500 g
Beilstein 3768231
Merck Index 14,5343
Solubility Information Soluble in water (warm) (0.1g/mL),and ethanol (very slightly soluble). Insoluble in chloroform,and ether.
Formula Weight 360.32 (342.30 Anhydrous)
Grade ACS
Density 1.525 g/mL
Chemical Name or Material α-D-Lactose monohydrate, For analysis ACS
EINECSNumber 200-559-2
RTECSNumber OD9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only