accessibility menu, dialog, popup

UserName

H2OHO(R)O(S)O(S)(R)HO(R)OH(S)OHO(R)HO(R)OH(S)HO(R)HO

CAS RN 5989-81-1

IUPAC Name: (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
Synonyms: α-lactose hydrate
Molecular Weight (g/mol): 360.31
Molecular Formula: C12H24O12
InChi Key: WSVLPVUVIUVCRA-KPKNDVKVSA-N
SMILES: O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)