Learn More
D-Gulonic acid-1,4-lactone, 98%
CAS: 6322-07-2 | C6H10O6 | 178.14 g/mol
Supplier: Thermo Scientific Chemicals J6698714
Description
D-Gulono-1,4-lactone is a hexanoic acid used as a starting material in EPC syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Gulono-1,4-lactone is a hexanoic acid used as a starting material in EPC syntheses.
Solubility
Soluble in water
Notes
Store at room temperature. Incompatible with heat, oxidizing agents. Ensure good ventilation.
Specifications
| D-Gulonic acid-1,4-lactone | |
| 182°C to 188°C | |
| C6H10O6 | |
| 25 g | |
| d-gulono-1,4-lactone, d---gulonic acid gamma-lactone, gulonolactone, d-gulonic acid gamma-lactone, gulono-1,4-lactone, d-gulonic acid-1,4-lactone, 3r,4s,5s-5-r-1,2-dihydroxyethyl-3,4-dihydroxydihydrofuran-2 3h-one, 3r,4s,5s-5-1r-1,2-dihydroxyethyl-3,4-dihydroxyoxolan-2-one, d-gulono-gamma-lactone, gulonic acid-1,4-lactone | |
| SXZYCXMUPBBULW-UHFFFAOYNA-N | |
| (3R,4S,5S)-5-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one | |
| 165105 | |
| 98% |
| 6322-07-2 | |
| White | |
| MFCD00005391 | |
| 83012 | |
| Soluble in water | |
| C(C(C1C(C(C(=O)O1)O)O)O)O | |
| 178.14 | |
| 178.14 | |
| Powder |
Chemical Identifiers
| 6322-07-2 | |
| MFCD00005391 |
Safety and Handling
EINECSNumber : 228-678-5
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only