missing translation for 'onlineSavingsMsg'
Learn More

D-Gulonic acid-1,4-lactone, 98%

Catalog No. p-7043302
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6698706 5 g
AAJ6698714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6698706 Supplier Thermo Scientific Chemicals Supplier No. J6698706
Only null left

CAS: 6322-07-2 | C6H10O6 | 178.14 g/mol

D-Gulono-1,4-lactone is a hexanoic acid used as a starting material in EPC syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Gulono-1,4-lactone is a hexanoic acid used as a starting material in EPC syntheses.

Solubility
Soluble in water

Notes
Store at room temperature. Incompatible with heat, oxidizing agents. Ensure good ventilation.

Chemical Identifiers

CAS 6322-07-2
MDL Number MFCD00005391

Specifications

Melting Point 182°C to 188°C
Color White
Molecular Formula C5H2Br2O2S
Quantity 5 g
Beilstein 83012
Synonym 4,5-dibromo-2-thiophenecarboxylic acid, 4,5-dibromothenoic acid, 4,5-dibromo-thiophene-2-carboxylic acid, pubchem7747, acmc-20a4op, maybridge1_002274, 4-dimethylaminoethyl benzoate, 4,5-dibromo-2-thiophencarboxylic acid, 2,3-dibromothiophene-5-carboxylic acid, 2-thiophenecarboxylicacid, 4,5-dibromo
Solubility Information Soluble in water
InChI Key USFXRYVNRUMABJ-UHFFFAOYSA-N
SMILES C1=C(SC(=C1Br)Br)C(=O)O
IUPAC Name 4,5-dibromothiophene-2-carboxylic acid
Molecular Weight (g/mol) 285.94
PubChem CID 80611
Formula Weight 178.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Gulonic acid-1,4-lactone
Show More Show Less
EINECSNumber 228-678-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only