accessibility menu, dialog, popup

UserName

CAS RN 6322-07-2

IUPAC Name: 5-(1,2-dihydroxyethyl)-3,4-dihydroxyoxolan-2-one
Synonyms: gulono-1,4-lactone
Molecular Weight (g/mol): 178.14
Molecular Formula: C6H10O6
InChi Key: SXZYCXMUPBBULW-UHFFFAOYNA-N
SMILES: OCC(O)C1OC(=O)C(O)C1O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)