missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucose, ACS Reagent, anhydrous

CAS: 50-99-7 | C6H12O6 | 180.156 g/mol

Supplier:  Thermo Scientific Chemicals 410950010

 View more versions of this product

Catalog No. AC410950010


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D(+)-Glucose
50-99-7
White
0.2% max. (105°C, 6 hrs)
Plastic Bottle
MFCD00063684
31, 83
+ 52.75 (25.00°C c=10,H2O,NH4OH)
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
GZCGUPFRVQAUEE-SLPGGIOYSA-N
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
107526
180.16
Crystalline Powder
Anhydrous, For analysis ACS
146.0°C
0.02% max.
Authentic
C6H12O6
1 kg
15, 4495
+ 52.75
0.005% max. Insoluble Matter
C(C(C(C(C(C=O)O)O)O)O)O
180.156
CHEBI:42758
ACS Reagent

Chemical Identifiers

50-99-7
GZCGUPFRVQAUEE-SLPGGIOYSA-N
107526
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
MFCD00063684
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
CHEBI:42758
C(C(C(C(C(C=O)O)O)O)O)O
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only