missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucose, ACS Reagent, anhydrous

Catalog No. p-3758068
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC410955000 500 g Plastic Bottle
AC410950010 1 kg Plastic Bottle
AC410950050 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC410955000 Supplier Thermo Scientific Chemicals Supplier No. 410955000
Only null left

CAS: 50-99-7 | C6H12O6 | 180.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-99-7
MDL Number MFCD00063684
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 146.0°C
Color White
Ignition Residue 0.02% max.
Loss on Drying 0.2% max. (105°C, 6 hrs)
Infrared Spectrum Authentic
Packaging Plastic Bottle
Molecular Formula C6H12O6
Quantity 500 g
Beilstein 31, 83
Merck Index 15, 4495
Specific Rotation Condition + 52.75 (25.00°C c=10,H2O,NH4OH)
Specific Rotation + 52.75
Solubility Information 0.005% max. Insoluble Matter
Molecular Weight (g/mol) 180.156
Formula Weight 180.16
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Glucose, Anhydrous, For analysis ACS
Show More Show Less

RUO – Research Use Only