missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Glucose-6-phosphate disodium salt hydrate, 98%

Catalog No. AAJ6602306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6602306 5 g
AAJ6602303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6602306 Supplier Thermo Scientific Chemicals Supplier No. J6602306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3671-99-6 | C6H11Na2O9P | 304.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3671-99-6
Molecular Formula C6H11Na2O9P
Molecular Weight (g/mol) 304.10
MDL Number MFCD00136037
InChI Key CHIBNKGHYAQTQY-OGPXQWFKNA-L
Synonym d-glucose-6-phosphate disodium salt, sodium 2r,3r,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexyl phosphate, sodium glucose 6-phosphate, disodium d-glucose 6-phosphate, glucose 6-phosphate sodium salt, glucose-6-phosphate disodium, disodium d-glucose-6-phosphate, d-glucose 6-disodium phosphate, d-glucose 6-phosphoric acid disodium salt, disodium 2r,3s,4r,5r-2,3,4,5-tetrahydroxy-6-phosphonooxy hexanal
PubChem CID 18594434
IUPAC Name disodium;[(2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl] phosphate
SMILES [Na+].[Na+].O[C@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in water
Formula Weight 304.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Glucose-6-phosphate disodium salt
EINECSNumber 222-938-1
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only