accessibility menu, dialog, popup

UserName

CAS RN 3671-99-6

(S)(R)(S)OHO(S)(R)OHHOHOOPOOONaNa

CAS RN 3671-99-6

IUPAC Name: disodium [(2R,3S,4S,5R,6S)-3,4,5,6-tetrahydroxyoxan-2-yl]methyl phosphate
Synonyms: disodium α-D-glucose 6-phosphate(2-)
Molecular Weight (g/mol): 304.1
Molecular Formula: C6H11Na2O9P
InChi Key: CHIBNKGHYAQTQY-OGPXQWFKNA-L
SMILES: [Na+].[Na+].O[C@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)[C@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results