missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Glucono-1,5-lactone 98.0+%, TCI America™
$32.72 - $67.38
Chemical Identifiers
| CAS | 90-80-2 |
|---|---|
| Molecular Formula | C6H10O6 |
| Molecular Weight (g/mol) | 178.14 |
| MDL Number | MFCD00006647 |
| InChI Key | PHOQVHQSTUBQQK-UHFFFAOYNA-N |
| Synonym | gluconolactone, delta-gluconolactone, d-glucono-1,5-lactone, d-gluconolactone, 1,5-gluconolactone, d-gluconic acid lactone, gluconic acid lactone, glucono delta-lactone, 1,5-d-gluconolactone, gluconic lactone |
| PubChem CID | 7027 |
| ChEBI | CHEBI:16217 |
| IUPAC Name | 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one |
| SMILES | OCC1OC(=O)C(O)C(O)C1O |
Chemical Identifiers
| 90-80-2 | |
| 178.14 | |
| PHOQVHQSTUBQQK-UHFFFAOYNA-N | |
| 7027 | |
| 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one |
| C6H10O6 | |
| MFCD00006647 | |
| gluconolactone, delta-gluconolactone, d-glucono-1,5-lactone, d-gluconolactone, 1,5-gluconolactone, d-gluconic acid lactone, gluconic acid lactone, glucono delta-lactone, 1,5-d-gluconolactone, gluconic lactone | |
| CHEBI:16217 | |
| OCC1OC(=O)C(O)C(O)C1O |
Specifications
| 90-80-2 | |
| White | |
| MFCD00006647 | |
| gluconolactone, delta-gluconolactone, d-glucono-1,5-lactone, d-gluconolactone, 1,5-gluconolactone, d-gluconic acid lactone, gluconic acid lactone, glucono delta-lactone, 1,5-d-gluconolactone, gluconic lactone | |
| OCC1OC(=O)C(O)C(O)C1O | |
| 178.14 | |
| CHEBI:16217 | |
| ≥98.0% (T) | |
| D-(+)-Glucono-1,5-lactone |
| 153°C | |
| C6H10O6 | |
| 25 g | |
| PHOQVHQSTUBQQK-UHFFFAOYNA-N | |
| 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one | |
| 7027 | |
| 178.14 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3668&(8)-0524
RTECSNumber : LZ5184000
TSCA : Yes