missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucono-1,5-lactone 98.0+%, TCI America™

Catalog No. G003925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G003925G 25 g
G0039500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G003925G Supplier TCI America Supplier No. G003925G

Chemical Identifiers

CAS 90-80-2
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00006647
InChI Key PHOQVHQSTUBQQK-UHFFFAOYNA-N
Synonym gluconolactone, delta-gluconolactone, d-glucono-1,5-lactone, d-gluconolactone, 1,5-gluconolactone, d-gluconic acid lactone, gluconic acid lactone, glucono delta-lactone, 1,5-d-gluconolactone, gluconic lactone
PubChem CID 7027
ChEBI CHEBI:16217
IUPAC Name 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
SMILES OCC1OC(=O)C(O)C(O)C1O

Specifications

Melting Point 153°C
Color White
Quantity 25 g
Formula Weight 178.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Glucono-1,5-lactone
EINECSNumber (5)-3668&(8)-0524
RTECSNumber LZ5184000
TSCA Yes