Learn More
SHOWA™ 910 Stainless Steel/HPPE Cut-Resistant Gloves
Flexible and lightweight gloves are available in uncoated or PVC coated styles and are ideal for light-duty applications.
$10.28 - $12.45
Specifications
| Certifications/Compliance (Europe) | CE |
|---|---|
| Construction | Seamless Knit |
| Color | White |
| Cut Resistance Rating | ANSI A4 |
| Hand | Ambidextrous |
| Catalog Number | Mfr. No. | Size | Coating Material | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Coating Material | Price | Quantity | ||||
|
19048577A
|
SHOWA™
910C06 |
6 | PVC Dots |
Each for $12.45
Case of 12 Each for $144.84
|
|
||||
|
19-048577B
|
SHOWA™
910C07 |
7 | PVC Dots |
Each for $12.45
Case of 12 Each for $144.84
|
|
||||
|
19-048577C
|
SHOWA™
910C08 |
8 | PVC Dots |
Each for $12.45
Case of 12 Each for $144.84
|
|
||||
|
19-048577D
|
SHOWA™
910C09 |
9 | PVC Dots |
Each for $12.45
Case of 12 Each for $144.84
|
|
||||
|
19-048577E
|
SHOWA™
910C10 |
10 | PVC Dots |
Each for $12.45
Case of 12 Each for $144.84
|
|
||||
|
19048576A
|
SHOWA™
91006 |
6 | Uncoated |
Each for $10.28
Case of 12 Each for $119.52
|
|
||||
|
28-707-84
|
SHOWA™
910M08 |
Medium | Uncoated |
Each for $10.28
Case of 12 Each for $119.52
|
|
||||
|
28-707-85
|
SHOWA™
910L09 |
9 | Uncoated |
Each for $10.28
Case of 12 Each for $119.52
|
|
||||
|
28-707-86
|
SHOWA™
910XL10 |
10 | Uncoated |
Each for $10.28
Case of 12 Each for $119.52
|
|
||||
Description
- High-performance polyethylene engineered fiber for added strength, comfort and cut resistance
- Fully launderable and color coded for easy identification
- Gauge: 10
- Uncoated or PVC dotted style available
Specifications
| CE | |
| White | |
| Ambidextrous | |
| Cut-resistant Gloves | |
| Knit |
| Seamless Knit | |
| ANSI A4 | |
| Polyethylene | |
| EN 388:2X43DP | |
| 10 |