Learn More
SHOWA™ 910 Stainless Steel/HPPE Cut-Resistant Gloves
Flexible and lightweight gloves are available in uncoated or PVC coated styles and are ideal for light-duty applications.
Supplier:  SHOWA™ 910C07
 View more versions of this product
Catalog No. 19-048577B
Only null left
Explore available promotions
Description
Description
  • High-performance polyethylene engineered fiber for added strength, comfort and cut resistance
  • Fully launderable and color coded for easy identification
  • Gauge: 10
  • Uncoated or PVC dotted style available
Specifications
Specifications
CE
PVC Dots
ANSI A4
Polyethylene
7
Machine or compartment assembly, bottling, sheet Metal handling, construction, recycling, Glass handling, razor blades
10
Seamless Knit
White
Ambidextrous
Cut-resistant Gloves
EN 388:2X43DP
Knit
Product Suggestions
Product Suggestions