missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-(+)-Dextrose, (Crystalline), MP Biomedicals
Manufacturer: Mp Biomedicals 0290152105
Specifications
D-(+)-Dextrose | |
C6H12O6 | |
5kg | |
C(C(C(C(C(C=O)O)O)O)O)O | |
180.156 | |
CHEBI:42758 | |
Crystalline |
50-99-7 | |
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, aldehydo-d-glucose, anhydrous dextrose, d +-glucose, d-glucose in linear form, d-glucose, anhydrous, dextrose solution, dextrose, anhydrous, glucose, anhydrous, glucosteril | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal | |
107526 | |
180.2 |
Chemical Identifiers
50-99-7 | |
180.156 | |
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, aldehydo-d-glucose, anhydrous dextrose, d +-glucose, d-glucose in linear form, d-glucose, anhydrous, dextrose solution, dextrose, anhydrous, glucose, anhydrous, glucosteril | |
CHEBI:42758 | |
C(C(C(C(C(C=O)O)O)O)O)O |
C6H12O6 | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
107526 | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
Safety and Handling
Recommended Storage : Room Temperature