missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Cellobiose, 98+%

CAS: 528-50-7 | C12H22O11 | 342.30 g/mol

$56.93 - $443.95

Chemical Identifiers

CAS 528-50-7
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00136034
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
PubChem CID 57370084
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1455306
View Documents Promotion Details
Thermo Scientific Chemicals
A1455306
5 g
Each for $56.93
Only null left
 
AAA1455314
View Documents Promotion Details
Thermo Scientific Chemicals
A1455314
25 g
Each for $149.89
Only null left
 
AAA1455322
View Documents Promotion Details
Thermo Scientific Chemicals
A1455322
100 g
Each for $443.95
Only null left
 
Description

Description

D-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.

Solubility
Soluble in water (1 g/8ml). Insoluble in ether and ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

528-50-7
342.30
GUBGYTABKSRVRQ-UHFFFAOYNA-N
57370084
C12H22O11
MFCD00136034
d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

528-50-7
≥98%
MFCD00136034
14,1961
Soluble in water (1g/8ml). Insoluble in ether and ethanol.
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
(3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
57370084
≥98%
234°C (decomposition)
C12H22O11
93795
d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
GUBGYTABKSRVRQ-UHFFFAOYNA-N
+34° (c=10 in Water, 15h)
342.30
342.3
D-(+)-Cellobiose
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-436-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only