missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Cellobiose, 98+%

CAS: 528-50-7 | C12H22O11 | 342.30 g/mol

Supplier:  Thermo Scientific Chemicals A1455314

Quantity 25 g

 View more versions of this product

Catalog No. AAA1455314


Only null left

Description

Description

D-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Cellobiose is used to differentiate bacteria based on carbohydrate fermentation abilities. It is a β-glucosidase substrate. Also used as as a chemical raw material and for foods, as a low-sweetness sugar.

Solubility
Soluble in water (1 g/8ml). Insoluble in ether and ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

528-50-7
342.30
GUBGYTABKSRVRQ-UHFFFAOYNA-N
57370084
C12H22O11
MFCD00136034
d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

D-(+)-Cellobiose
234°C (decomposition)
C12H22O11
MFCD00136034
14,1961
Soluble in water (1g/8ml). Insoluble in ether and ethanol.
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
342.30
342.3
528-50-7
≥98%
25 g
93795
d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
GUBGYTABKSRVRQ-UHFFFAOYNA-N
(3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
57370084
≥98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-436-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions