missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D and C Green No. 5, Spectrum™ Chemical
D1119, 4403-90-1, C28H20N2O8S2Na2
$275.43 - $2,619.46
Chemical Identifiers
| CAS | 4403-90-1 |
|---|---|
| Molecular Formula | C28H20N2Na2O8S2 |
| Molecular Weight (g/mol) | 622.57 |
| InChI Key | FPAYXBWMYIMERV-UHFFFAOYSA-L |
| IUPAC Name | disodium 5-methyl-2-({4-[(4-methyl-2-sulfonatophenyl)amino]-9,10-dioxo-9,10-dihydroanthracen-1-yl}amino)benzene-1-sulfonate |
| SMILES | [Na+].[Na+].CC1=CC=C(NC2=CC=C(NC3=CC=C(C)C=C3S([O-])(=O)=O)C3=C2C(=O)C2=CC=CC=C2C3=O)C(=C1)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18601967
|
Spectrum Chemical Mfg Cor
D111925GM |
25 g | Amber Glass Bottle |
N/A
|
|
||||
|
18601968
|
Spectrum Chemical Mfg Cor
D1119100GM |
100 g | Amber Glass Bottle |
N/A
|
|
||||
|
18601969
|
Spectrum Chemical Mfg Cor
D1119500GM |
500 g | Amber Glass Bottle |
N/A
|
|
||||
Description
Spectrum™ Chemical D and C Green No. 5Chemical Identifiers
| 4403-90-1 | |
| 622.57 | |
| disodium 5-methyl-2-({4-[(4-methyl-2-sulfonatophenyl)amino]-9,10-dioxo-9,10-dihydroanthracen-1-yl}amino)benzene-1-sulfonate |
| C28H20N2Na2O8S2 | |
| FPAYXBWMYIMERV-UHFFFAOYSA-L | |
| [Na+].[Na+].CC1=CC=C(NC2=CC=C(NC3=CC=C(C)C=C3S([O-])(=O)=O)C3=C2C(=O)C2=CC=CC=C2C3=O)C(=C1)S([O-])(=O)=O |
Specifications
| 4403-90-1 | |
| C28H20N2Na2O8S2 | |
| [Na+].[Na+].CC1=CC=C(NC2=CC=C(NC3=CC=C(C)C=C3S([O-])(=O)=O)C3=C2C(=O)C2=CC=CC=C2C3=O)C(=C1)S([O-])(=O)=O | |
| 622.57 | |
| Amber Glass Bottle | |
| 25 g |
| 100% | |
| FPAYXBWMYIMERV-UHFFFAOYSA-L | |
| disodium 5-methyl-2-({4-[(4-methyl-2-sulfonatophenyl)amino]-9,10-dioxo-9,10-dihydroanthracen-1-yl}amino)benzene-1-sulfonate | |
| Reagent | |
| Blue/Green | |
| D and C Green No. 5 |