accessibility menu, dialog, popup

UserName

CAS RN 4403-90-1

NaNaH3CNHNHH3CSOOOOOSOOO

CAS RN 4403-90-1

IUPAC Name: disodium 5-methyl-2-({4-[(4-methyl-2-sulfonatophenyl)amino]-9,10-dioxo-9,10-dihydroanthracen-1-yl}amino)benzene-1-sulfonate
Synonyms: alizarin cyanine green F
Molecular Weight (g/mol): 622.57
Molecular Formula: C28H20N2Na2O8S2
InChi Key: FPAYXBWMYIMERV-UHFFFAOYSA-L
SMILES: [Na+].[Na+].CC1=CC=C(NC2=CC=C(NC3=CC=C(C)C=C3S([O-])(=O)=O)C3=C2C(=O)C2=CC=CC=C2C3=O)C(=C1)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Color 
  • (3)
Grade 
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results