Learn More
D-beta-Prolinol, 95%
CAS: 110013-18-8 | C5H11NO | 101.149 g/mol
$191.51 - $661.56
Chemical Identifiers
| CAS | 110013-18-8 |
|---|---|
| Molecular Formula | C5H11NO |
| Molecular Weight (g/mol) | 101.149 |
| MDL Number | MFCD09607969 |
| InChI Key | QOTUIIJRVXKSJU-RXMQYKEDSA-N |
| Synonym | r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-yl methanol, r-3-hydroxymethyl pyrrolidine, d-beta-prolinol, 3-pyrrolidinemethanol, 3r, r-3-pyrrolidin-methanol, r-3-pyrrolidinemethanol, 3r-3-pyrrolidinemethanol |
| PubChem CID | 7446913 |
| IUPAC Name | [(3R)-pyrrolidin-3-yl]methanol |
| SMILES | C1CNCC1CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH57871MD
|
N/A | 250 mg |
Each for $191.51
|
|
|||||
|
AAH5787103
|
N/A | 1 g |
Each for $661.56
|
|
|||||
Description
It is an intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an intermediate in organic syntheses.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Store at room temperature.
Chemical Identifiers
| 110013-18-8 | |
| 101.149 | |
| QOTUIIJRVXKSJU-RXMQYKEDSA-N | |
| 7446913 | |
| C1CNCC1CO |
| C5H11NO | |
| MFCD09607969 | |
| r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-yl methanol, r-3-hydroxymethyl pyrrolidine, d-beta-prolinol, 3-pyrrolidinemethanol, 3r, r-3-pyrrolidin-methanol, r-3-pyrrolidinemethanol, 3r-3-pyrrolidinemethanol | |
| [(3R)-pyrrolidin-3-yl]methanol |
Specifications
| 110013-18-8 | |
| 173°C to 177°C | |
| C5H11NO | |
| r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-yl methanol, r-3-hydroxymethyl pyrrolidine, d-beta-prolinol, 3-pyrrolidinemethanol, 3r, r-3-pyrrolidin-methanol, r-3-pyrrolidinemethanol, 3r-3-pyrrolidinemethanol | |
| QOTUIIJRVXKSJU-RXMQYKEDSA-N | |
| [(3R)-pyrrolidin-3-yl]methanol | |
| 7446913 | |
| 95% |
| 0.979 | |
| Hygroscopic | |
| MFCD09607969 | |
| Slightly soluble in water. | |
| C1CNCC1CO | |
| 101.149 | |
| 101.15 | |
| D-beta-Prolinol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only