missing translation for 'onlineSavingsMsg'
Learn More

D-beta-Prolinol, 95%

CAS: 110013-18-8 | C5H11NO | 101.149 g/mol

Supplier:  Thermo Scientific Chemicals H5787103

Quantity 1 g

 View more versions of this product

Catalog No. AAH5787103


Only null left

Description

Description

It is an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Store at room temperature.
Specifications

Chemical Identifiers

110013-18-8
101.149
QOTUIIJRVXKSJU-RXMQYKEDSA-N
7446913
C1CNCC1CO
C5H11NO
MFCD09607969
r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-yl methanol, r-3-hydroxymethyl pyrrolidine, d-beta-prolinol, 3-pyrrolidinemethanol, 3r, r-3-pyrrolidin-methanol, r-3-pyrrolidinemethanol, 3r-3-pyrrolidinemethanol
[(3R)-pyrrolidin-3-yl]methanol

Specifications

D-beta-Prolinol
0.979
1 g
C5H11NO
r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-yl methanol, r-3-hydroxymethyl pyrrolidine, d-beta-prolinol, 3-pyrrolidinemethanol, 3r, r-3-pyrrolidin-methanol, r-3-pyrrolidinemethanol, 3r-3-pyrrolidinemethanol
QOTUIIJRVXKSJU-RXMQYKEDSA-N
[(3R)-pyrrolidin-3-yl]methanol
7446913
95%
110013-18-8
173°C to 177°C
Hygroscopic
MFCD09607969
Slightly soluble in water.
C1CNCC1CO
101.149
101.15
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions