missing translation for 'onlineSavingsMsg'
Learn More

D-beta-Prolinol, 95%

Catalog No. AAH57871MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57871MD 250 mg
AAH5787103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57871MD Supplier Thermo Scientific Chemicals Supplier No. H57871MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110013-18-8 | C5H11NO | 101.149 g/mol

It is an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Store at room temperature.

Chemical Identifiers

CAS 110013-18-8
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD09607969
InChI Key QOTUIIJRVXKSJU-RXMQYKEDSA-N
Synonym r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-ylmethanol, 3r-pyrrolidin-3-yl methanol, r-3-hydroxymethyl pyrrolidine, d-beta-prolinol, 3-pyrrolidinemethanol, 3r, r-3-pyrrolidin-methanol, r-3-pyrrolidinemethanol, 3r-3-pyrrolidinemethanol
PubChem CID 7446913
IUPAC Name [(3R)-pyrrolidin-3-yl]methanol
SMILES C1CNCC1CO

Specifications

Density 0.979
Boiling Point 173°C to 177°C
Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 101.15
Percent Purity 95%
Chemical Name or Material D-beta-Prolinol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only