missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Aspartic acid 1-methyl ester, 98%
CAS: 65414-78-0 | C5H9NO4 | 147.13 g/mol
$145.47 - $563.14
Chemical Identifiers
| CAS | 65414-78-0 |
|---|---|
| Molecular Formula | C5H9NO4 |
| Molecular Weight (g/mol) | 147.13 |
| MDL Number | MFCD03424227 |
| InChI Key | SWWBMHIMADRNIK-GSVOUGTGSA-N |
| Synonym | h-d-asp-ome, r-3-amino-4-methoxy-4-oxobutanoic acid, d-asp-ome, d-aspartic acid 1-methyl ester, d-aspartic acid,1-methyl ester, 3r-3-amino-4-methoxy-4-oxobutanoic acid |
| PubChem CID | 7020023 |
| IUPAC Name | (3R)-3-amino-4-methoxy-4-oxobutanoic acid |
| SMILES | COC(=O)C(CC(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6365806
|
Thermo Scientific Chemicals
H6365806 |
5 g |
Each for $145.47
|
|
|||||
|
AAH6365814
|
Thermo Scientific Chemicals
H6365814 |
25 g |
Each for $563.14
|
|
|||||
Description
D-Aspartic acid 1-methyl ester is used as a intermediate for pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Aspartic acid 1-methyl ester is used as a intermediate for pharmaceutical and chemical research.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
| 65414-78-0 | |
| 147.13 | |
| SWWBMHIMADRNIK-GSVOUGTGSA-N | |
| 7020023 | |
| COC(=O)C(CC(=O)O)N |
| C5H9NO4 | |
| MFCD03424227 | |
| h-d-asp-ome, r-3-amino-4-methoxy-4-oxobutanoic acid, d-asp-ome, d-aspartic acid 1-methyl ester, d-aspartic acid,1-methyl ester, 3r-3-amino-4-methoxy-4-oxobutanoic acid | |
| (3R)-3-amino-4-methoxy-4-oxobutanoic acid |
Specifications
| 65414-78-0 | |
| MFCD03424227 | |
| SWWBMHIMADRNIK-GSVOUGTGSA-N | |
| (3R)-3-amino-4-methoxy-4-oxobutanoic acid | |
| 147.13 | |
| 147.13 | |
| Solid |
| C5H9NO4 | |
| h-d-asp-ome, r-3-amino-4-methoxy-4-oxobutanoic acid, d-asp-ome, d-aspartic acid 1-methyl ester, d-aspartic acid,1-methyl ester, 3r-3-amino-4-methoxy-4-oxobutanoic acid | |
| COC(=O)C(CC(=O)O)N | |
| 5 g | |
| 7020023 | |
| 98% | |
| D-Aspartic acid 1-methyl ester |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only