CAS RN 65414-78-0
IUPAC Name:
3-amino-4-methoxy-4-oxobutanoic acid
Synonyms:
3-amino-4-methoxy-4-oxobutanoic acid
Molecular Weight (g/mol):
147.13
Molecular Formula:
C5H9NO4
InChi Key:
SWWBMHIMADRNIK-UHFFFAOYNA-N
SMILES:
COC(=O)C(N)CC(O)=O
1
–
1
of
1
results
1
Promotions Available
D-Aspartic acid 1-methyl ester, 98%
D-Aspartic acid 1-methyl ester, 98%
CAS: 65414-78-0 Molecular Formula: C5H9NO4 Molecular Weight (g/mol): 147.13 MDL Number: MFCD03424227 InChI Key: SWWBMHIMADRNIK-GSVOUGTGSA-N Synonym: h-d-asp-ome,r-3-amino-4-methoxy-4-oxobutanoic acid,d-asp-ome,d-aspartic acid 1-methyl ester,d-aspartic acid,1-methyl ester,3r-3-amino-4-methoxy-4-oxobutanoic acid PubChem CID: 7020023 IUPAC Name: (3R)-3-amino-4-methoxy-4-oxobutanoic acid SMILES: COC(=O)C(CC(=O)O)N