Learn More
D-Aspartic acid 1-methyl ester, 98%
CAS: 65414-78-0 | C5H9NO4 | 147.13 g/mol
Supplier: Thermo Scientific Chemicals H6365814
Description
D-Aspartic acid 1-methyl ester is used as a intermediate for pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Aspartic acid 1-methyl ester is used as a intermediate for pharmaceutical and chemical research.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
| D-Aspartic acid 1-methyl ester | |
| C5H9NO4 | |
| h-d-asp-ome, r-3-amino-4-methoxy-4-oxobutanoic acid, d-asp-ome, d-aspartic acid 1-methyl ester, d-aspartic acid,1-methyl ester, 3r-3-amino-4-methoxy-4-oxobutanoic acid | |
| COC(=O)C(CC(=O)O)N | |
| 25 g | |
| 7020023 | |
| 98% |
| 65414-78-0 | |
| MFCD03424227 | |
| SWWBMHIMADRNIK-GSVOUGTGSA-N | |
| (3R)-3-amino-4-methoxy-4-oxobutanoic acid | |
| 147.13 | |
| 147.13 | |
| Solid |
Chemical Identifiers
| 65414-78-0 | |
| 147.13 | |
| SWWBMHIMADRNIK-GSVOUGTGSA-N | |
| 7020023 | |
| COC(=O)C(CC(=O)O)N |
| C5H9NO4 | |
| MFCD03424227 | |
| h-d-asp-ome, r-3-amino-4-methoxy-4-oxobutanoic acid, d-asp-ome, d-aspartic acid 1-methyl ester, d-aspartic acid,1-methyl ester, 3r-3-amino-4-methoxy-4-oxobutanoic acid | |
| (3R)-3-amino-4-methoxy-4-oxobutanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only