missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(-)-Arabinose 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 161°C |
| Quantity | 100 g |
| Formula Weight | 150.13 |
| Percent Purity | ≥99.0% (HPLC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | D-(-)-Arabinose |
Safety and Handling
| EINECSNumber | (2)-2470 |
| TSCA | Yes |