missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytosine, 99+%, Thermo Scientific™
$158.90 - $1,763.41
Chemical Identifiers
| CAS | 71-30-7 |
|---|---|
| Molecular Formula | C4H5N3O |
| Molecular Weight (g/mol) | 111.10 |
| MDL Number | MFCD00006034 |
| InChI Key | OPTASPLRGRRNAP-UHFFFAOYSA-N |
| Synonym | cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine |
| PubChem CID | 597 |
| ChEBI | CHEBI:16040 |
| IUPAC Name | 6-amino-1H-pyrimidin-2-one |
| SMILES | NC1=CC=NC(=O)N1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC161760050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC161760250
|
N/A | 25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC161761000
|
N/A | 100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C4H5N3O | |
| MFCD00006034 | |
| cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine | |
| CHEBI:16040 | |
| NC1=CC=NC(=O)N1 |
Specifications
| 71-30-7 | |
| Authentic | |
| C4H5N3O | |
| 24,314 | |
| cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine | |
| 98% min. (lambda max. 275nm,pH 1) molar absorptivity 10400+/-2%,on dry basis | |
| NC1=CC=NC(=O)N1 | |
| 111.10 | |
| CHEBI:16040 | |
| 99+% |
| 0.5% max. (105°C, 5 hrs) | |
| 99% min. (HPLC) | |
| MFCD00006034 | |
| 15,2792 | |
| Solubility in water: soluble. Other solubilities: soluble in alkalines and acids,slightly soluble in ethanol,chloroform,,insoluble in ether and hydrocarbons | |
| OPTASPLRGRRNAP-UHFFFAOYSA-N | |
| 6-amino-1H-pyrimidin-2-one | |
| 597 | |
| 111.1 | |
| Cytosine |
Safety and Handling
EINECSNumber : 200-749-5
RUO – Research Use Only