missing translation for 'onlineSavingsMsg'
Learn More

Cytosine, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 161760250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC161760250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

71-30-7
111.10
OPTASPLRGRRNAP-UHFFFAOYSA-N
597
6-amino-1H-pyrimidin-2-one
C4H5N3O
MFCD00006034
cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine
CHEBI:16040
NC1=CC=NC(=O)N1

Specifications

Cytosine
0.5% max. (105°C, 5 hrs)
99% min. (HPLC)
C4H5N3O
MFCD00006034
15,2792
Solubility in water: soluble. Other solubilities: soluble in alkalines and acids,slightly soluble in ethanol,chloroform,,insoluble in ether and hydrocarbons
OPTASPLRGRRNAP-UHFFFAOYSA-N
6-amino-1H-pyrimidin-2-one
597
111.1
71-30-7
Authentic
Glass bottle
25g
24,314
cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine
98% min. (lambda max. 275nm,pH 1) molar absorptivity 10400+/-2%,on dry basis
NC1=CC=NC(=O)N1
111.10
CHEBI:16040
99+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-749-5

SDS
Documents

Documents

Promotions

Promotions