missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 10 N Aqueous Solution, APHA, Spectrum™ Chemical

Catalog No. 18606545
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18606545 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606545 Supplier Spectrum Chemical Mfg Cor Supplier No. S3951LT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S-395, 1310-73-2

Spectrum™ Chemical Sodium Hydroxide, 10.0 N Aqueous Solution, APHA - As an APHA certified solution, this item meets the ASTM D1209 standards for use in water and waste water treatment facilities to determine the concentration of dissolved and particulate materials. Spectrum Chemical manufactured APHA grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 0.3, 0.7
Concentration 10 N ±0.5
Quantity 1 L
Grade APHA
Packaging Poly Bottle
Physical Form Chemical Solution
Chemical Name or Material Sodium Hydroxide Solution