missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome b-245/p22phox Mouse anti-Human, Clone: 44.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS2192100UG
Description
Cytochrome b-245/p22phox Monoclonal antibody specifically detects Cytochrome b-245/p22phox in Human samples. It is validated for Flow Cytometry, Western Blotting.Specifications
| Cytochrome b-245/p22phox | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, Western Blot | |
| 44.1 | |
| Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| Partially purified human neutrophil flavocytochrome b-245 light chain. | |
| 100 μg | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. |