missing translation for 'onlineSavingsMsg'
Learn More

Cytochrome b-245/p22phox Mouse anti-Human, Clone: 44.1, MilliporeSigma™

Mouse Monoclonal Antibody

Specifications

Antigen Cytochrome b-245/p22phox
Clone 44.1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS219225
View Documents Promotion Details
N/A 25 μg N/A
Only null left
 
MABS2192100
View Documents Promotion Details
N/A 100 μg N/A
Only null left
 
Description

Description

Cytochrome b-245/p22phox Monoclonal antibody specifically detects Cytochrome b-245/p22phox in Human samples. It is validated for Flow Cytometry, Western Blotting.
Specifications

Specifications

Cytochrome b-245/p22phox
Flow Cytometry, Western Blot
Unconjugated
Mouse
Human
Partially purified human neutrophil flavocytochrome b-245 light chain.
Protein G purified
44.1
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Primary
Stable for 1 year at 2-8°C from date of receipt.
Videos
SDS
Documents

Documents